VWR® Mini Stirrer, 120 V
Supplier: VWR International
10153-304 10153-314
10153-304EA 376.58 USD
10153-304 10153-314
VWR® Mini Stirrer, 120 V
Stirrers Magnetic Stirrers
Rugged, compact stirrers with an ergonomic design are ideal for educational labs and can stir up to 1000 mL of water.
  • Rugged 5x5" aluminum top plate
  • Speed range of 100 to 1200rpm
  • Built-in support rod holder
  • Five-Year Warranty on Parts and Labor
Durable 5x5" cast aluminum top plate will not chip or crack
The Auto Stirrers are ideal for applications that require repeatable results. These compact, powerful stirrers automatically start when a vessel is placed on the aluminum top plate (minimum weight of approximately 50 grams [1.75 oz.]). When theMore Product Information