Glass Vials, C&G Containers
Supplier: C&G Containers
LU23A000A01FZS LFV0403000 LV73A000A01BZS LV74H100B21BZS LV64H100B51BZS LAV040700005
71001-232CS 225.9 USD
71001-232 71001-234 71001-238 71001-240 71001-242 71001-244 71001-246 71001-248 71001-250 71001-252 71001-254 71001-256 71001-258 71001-260 71001-262 71001-264 71001-266 71001-268 71001-270 71001-276 71001-284 71001-288 71001-290 71001-292 71001-294 71001-296 71001-298 71001-300 71001-302 71001-304 71001-306 71001-310 71001-312 71001-314 71001-316 71001-318 71001-320 71001-322 71001-324 71001-326 71001-328 71001-330 71001-332 89137-128 89137-130 89137-132 89137-134 89136-722 89370-068 30617-868 30621-904 30621-906 30621-908 30621-912 30621-914 30621-916 30621-918 10146-996 10146-998 10496-920 10496-922 10496-926 10496-928 10787-058 75840-124
Glass Vials, C&G Containers
Vials
These glass vials from C&G Containers are designed primarily for the collection and testing of solid samples for Volatile Organic Analysis and Total Petroleum Hydrocarbons (TPH), and for the collection and testing of aqueous samples for Total Organic Carbon (TOC).
  • All glass vials are borosilicate Type 1 glass and use a 24-414 closure
Pre-cleaned and Certified products contain a Certificate of Analysis and Cleaning Protocol Form in the product case. Wash protocols for these vials include; A1 (Nitric acid wash) certified, B1 process certified, L3 (More Product Information