EcoSense ODO200 Optical DO and Temperature Meters, YSI
Supplier: YSI
606324 606325 606305 606329
10146-166EA 1408.46 USD
10146-166 10146-168 10146-170 10146-182
EcoSense ODO200 Optical DO and Temperature Meters, YSI
Electrochemistry Meters Oxygen Meters
The ODO200 is designed for quick, accurate results in an economical platform. The ability to measure DO optically along with temperature, in a simple, compact handheld allows the instrument to be used across multiple application sampling strategies in the field.
  • Automatic temperature compensation
  • 1-year instrument, cable and sensor cap warranty
  • 1-, 4- and 10-meter durable field cables available
  • Manual input for salinity and pressure compensation: 0.0 to 40.0 ppt (manual input)/ 500 to 1125 millibar (manual input)
  • Low battery indicator with 100 hour battery
More Product Information