Polar Imidazole HILIC Columns, Sepax Technologies
Supplier: Sepax Technologies
135333-2001 135333-4001 135333-4605 135335-2125 135335-4610 135335-10015 135339-4001 135335-2105 135335-4603 135335-4605 135333-2103 135333-2110 135339-21205 135333-2115 135333-4615 135335-2115 135335-4001 135335-4615 135335-7815 135335-2001 135333-2125 135333-4610 135333-4625 135335-2110 135335-7825 135335-4625 135333-2105 135333-4603 135335-2103 135339-7825 135331-2110 135331-2115 135331-2103 135331-2105 135331-4610 135331-4615 135331-4603 135331-4605 135339-4001C 135333-2001C 135333-4001C 135335-2001C 135335-4001C
101453-140EA 372.31 USD
101453-140 101453-152 101453-156 101453-174 101453-182 101453-192 101453-206 101453-168 101453-178 101453-180 101453-142 101453-146 101453-218 101453-148 101453-160 101453-172 101453-176 101453-184 101453-188 101453-164 101453-150 101453-158 101453-162 101453-170 101453-190 101453-186 101453-144 101453-154 101453-166 101453-208 103055-616 103055-618 103055-620 103055-622 103055-632 103055-634 103055-636 103055-638 103055-678 103058-472 103058-586 103058-588 103058-590
Polar Imidazole HILIC Columns, Sepax Technologies
Chromatography Columns
Polar-Imidazole phase is bonded with imidazole functional groups by utilizing highest purity and enhanced mechanical stability silica and pure bonding reagents. With a polar and N-containing ring structure, Polar-Pyridine phase is a weak base and adequate for many applications, such as pharmaceuticals, peptides, sugars, and nucleotides.
  • Spherical, high purity silica
  • Well controlled chemistry of polymeric monolayer
  • High column-to-column reproducibility
  • Analytical, semi-preparative, and preparative separation
  • HILIC phase for separation of acidic, neutral and
More Product Information