Precellys® CKMix50-R and CKMix50 Tissue Grinding Kits, Bertin
Supplier: BERTIN CORP
P000922-LYSK0-A.0 P000939-LYSK0-A.0 P000930-LYSK0-A.0
10144-520EA 377.73 USD
10144-520 10144-534 10144-564
Precellys® CKMix50-R and CKMix50 Tissue Grinding Kits, Bertin
Homogenizers Tissue Grinders
The CKMix50-R and CKMix50 Tissue Grinding Kits are designed for hard tissue grinding and homogenizing
  • Dnase/Rnase free
  • Inert 2.8 mm and 5.0 mm ceramic (zirconium oxide) beads
  • Autoclavable
  • Ideal for hard tissue grinding
The CKMix50 Tissue Grinding Kit contains 7 mL standard tubes, and is ideal for use with whole organs such as rat brain, liver, kidney, skin, whole iris-ciliary body pork, and dry samples such as pepper grains. All kits include 50 preps, ready-to-use with the Precellys®24, Precellys® Evolution, or Minilys devices. All tubes areMore Product Information