Medical Power Strips, Tripp Lite
Supplier: Tripp Lite
PS-615-HG PS-415-HGULTRA PS-606-HG PS-402-HG-OEM PS-410-HGOEMCC PS-415-HG PS-415-HG-OEM PS-615-HG-OEM SPS415HGULTRA PS625HG20AOEM PS615HG20AOEM PS607HG20AOEM PS-607-HG-OEM PS-407-HG-OEM PS-406-HGULTRA SPS406HGULTRA PS-602-HG
89079-128EA 192.96 USD
89079-128 89079-130 89079-132 10144-226 10144-228 10144-232 10144-234 10144-238 10144-444 76461-410 76461-412 76461-414 76461-416 76461-418 76461-420 76461-422 76461-424 76461-426 76461-428
Medical Power Strips, Tripp Lite
Electrical Sockets
Medical-grade power strips designed and manufactured in accordance with Article 517 and certified to meet UL standards.
  • Power distribution anywhere in any medical facility
  • Exclusive, patented antimicrobial protection
  • Versatile medical-grade solution
  • Premium safety features
  • Ready to use right out of the box
The Medical-Grade Power Strip supplies power to movable plug-connected components mounted to a rack, table or pedestal in operating rooms, patient-care vicinities, therapy rooms, imaging environments and administrative areas. Compliant with UL 1363AMore Product Information