[RuCl(p-cymene)((S)-dm-segphos®)]Cl

Supplier: STREM CHEMICALS INC MS
Warning

Synonyms: Chloro{(S)-(-)-5,5'-bis[di(3,5-xylyl)phosphino]-4,4'-bi-1,3-benzodioxole}(p-cymene)ruthenium(II) chloride [RuCl(p-cymene)((S)-dm-segphos®)]Cl, Chloro[(S)-(-)-5,5'-bis[di(3,5-xylyl)phosphino]-4,4'-bi-1,3-benzodioxole](p-cymene)ruthenium(II) Chloride

44-0099-250MG
101418-862EA 90.58 USD
101418-862
[RuCl(p-cymene)((S)-dm-segphos®)]Cl
[RuCl(p-cymene)((S)-dm-segphos®)]Cl

SEGPHOS

Formula: C₅₆H₅₈Cl₂O₄P₂Ru
MW: 1028.99 g/mol
Melting Pt: >100 °C (dec.)
MDL Number: MFCD09753018
CAS Number: 944451-31-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR