Graphite, stripper films 0.75 x 0.75 in (19 x 19' mm), pyrolytic

Supplier: Electron Microscopy Sciences

76040-10 76040-05
101413-660EA 1406.5 USD
101413-660 101413-658
Graphite, stripper films 0.75 x 0.75 in (19 x 19' mm), pyrolytic
Graphite

Smooth pyrolytic graphite stripper film is used in radiation sources, nuclear particle high energy density research, and medical diagnostic imaging instruments Pyrolytic graphite film offers major customer benefits relative to alternative materials such as amorphous carbon film or polycrystalline graphite films.


  • Stable at high temperature and vacuum
  • Easy to flex due to inherent higher flexural modulus
  • Erodes very slowly


These result in higher strength than alternative materials and films last 5 times longer than alternative materials. The foils can be customized up to 1.5" wide and 3 inches long from 5µm and up thicknesses.

Formula: C
MDL Number: MFCD00144065
CAS Number: 7782-42-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR