Sure Shot® Spray-Safe Atomizer, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
64186-20 64186-22 64189 64187
101412-896EA 98.63 USD
101412-896 101412-898 101412-892 101412-904 101412-900
Sure Shot® Spray-Safe Atomizer, Electron Microscopy Sciences
Atomizers
Sure Shot® pressure sprayers are available in 16, 8 and 7 oz. capacities.
  • Plated aluminum
  • Refillable and rechargeable
  • Working pressure: 80 to 145 psi
  • Measures are the same as aerosol cans (~2³/₄" Ø)
  • Liquid spray lighter than #10 motor oil
All sprayers come standard with Viton® seals, these seals are compatible with most products.