Crystal violet 1% in aqueous solution for Gram staining

Supplier: Electron Microscopy Sciences

Synonyms: Methylrosanilide chloride, Basic violet 3, Viocid, N-(4-(Bis(4-(dimethylamino)phenyl)methylene)cyclohexa-2,5-dien-1-ylidene)-N-methylmethanamonium chloride, Violet crystal, Methyl violet 10B, Dimethyl((di(4-(dimethylamino)phenyl)methylen)-2,5-cyclohexadien-1-ylidene)ammonium chloride, CI 42555, 4-[4,4'-Bis(dimethylamino) benzhydrylidene]cyclohexa-2,5-dien-1-ylidene]dimethylammonium chloride, Hexamethyl violet, Hexamethyl-p-rosaniline chloride, Vianin, Gentian violet

26105-01
101411-028EA 50.02 USD
101411-028
Crystal violet 1% in aqueous solution for Gram staining
Crystal Violet

Reagent for Brown and Brenn Method for Gram Positive and Gram Negative Bacteria


For bacteria stain: Gram + / Gram – bacteria and filaments of nocardia, etc


Stain results - Gram + Bacteria, Nocardia and Actinomyces Filaments: Blue, Gram – Bacteria, Nuclei: Red, Additional tissue elements: Yellow

Formula: C₂₅H₃₀ClN₃
MDL Number: MFCD00011750
CAS Number: 548-62-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR