Congo red 1% in aqueous solution

Supplier: Electron Microscopy Sciences
Danger

Synonyms: Disodium 3,3'-[[1,1'-biphenyl]-4,4'-diylbis(azo)]bis(4-aminonaphthalene-1-sulphonate), Sodium 3,3'-([1,1'-biphenyl]-4,4'-diylbis(diazene-2,1-diyl))bis(4-aminonaphthalene-1-sulphonate), Disodium 3,3'-[[1,1'-biphenyl]-4,4'-diylbis(azo)]bis(4-aminonaphthalene-1-sulfonate), CI 22120, Sodium 3,3'-([1,1'-biphenyl]-4,4'-diylbis(diazene-2,1-diyl))bis(4-aminonaphthalene-1-sulfonate), Direct Red 28

26090-25
101411-008EA 89.2 USD
101411-008
Congo red 1% in aqueous solution
Congo Red

  • Premixed ready-to-use; saves time and effort
  • Consistently uniform results are obtained
  • Packaged and prepared in convenient and precise quantities

Formula: C₃₂H₂₂N₆Na₂O₆S₂
MDL Number: MFCD00004028
CAS Number: 573-58-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR