Sodium thiosulfate 1% aqueous solution

Supplier: Electron Microscopy Sciences

Synonyms: di-Sodium thiosulfate, Sodiumthiosulphate, Sodium hyposulphite, Antichlor, di-Sodium thiosulphate, Sodiumthiosulfate, Sodium hyposulfite

21360-1
101410-812EA 56.98 USD
101410-812
Sodium thiosulfate 1% aqueous solution
Sodium thiosulphate

Sodium Thiosulfate solution Electron Microscopy Sciences is available in concentrations of 1.0% aqueous, 2.0% aqueous, and 10.0% aqueous.

Formula: Na₂S₂O₃
MW: 158.11 g/mol
Boiling Pt: 100
Melting Pt: 48
MDL Number: MFCD00003499
CAS Number: 7772-98-7
Merck Index: 12,08844
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=7c5803a1f089fdb2115119e13591b0f0dc54c2bc&vid=469aa0982e4240b7edca91051d4275aab09cdc05&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR