Picric acid saturated in aqueous solution

Supplier: Electron Microscopy Sciences

Synonyms: 2,4,6-Trinitrophenol, TNP

19552
101410-760EA 92.09 USD
101410-760
Picric acid saturated in aqueous solution
Picric acid
Formula: C₆H₃N₃O₇
MDL Number: MFCD00007102
CAS Number: 88-89-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=66df51b06177998b02c071534cebb6fc2461ef6c&vid=b62d8cb018b42a45f5b649fdb92e63f64cfaaf87&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR