Ethanol denatured SDA 200 Proof

Supplier: Decon Labs
Danger

Synonyms: Ethyl alcohol denatured

2955M
10141-264EA 2466.67 USD
10141-264
Ethanol denatured SDA 200 Proof
Ethanol denatured

Decon’s SDA 23A Ethanol, 200 proof is a specially denatured alcohol formulation used for research, healthcare, industrial, or production applications.



Decon’s SDA 23A Ethanol, 200 proof is a specially denatured alcohol formulation used for research, healthcare, industrial, or production applications. While specially denatured alcohol is exempt from Federal Excise Tax, please note that like pure ethanol, Decon’s SDA 23A ethanol, 200 Proof is regulated by the US ATF/TTB. To purchase you must have an Industrial Alcohol User Permit from the AFT/TTB. To apply for this permit visit www.ttb.gov and search for “Form 5150.22” - which is the application for the Industrial Alcohol User Permit.



This product offers high purity and reliability for various laboratory and manufacturing needs, providing consistent performance for cleaning, sanitization, and chemical processing while meeting stringent industry standards.



Packaging:

55 Gallon Drum


Formula: H₃CCH₂OH
MW: 46.07 g/mol
MDL Number: MFCD00003568
CAS Number: 64-17-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR