Anti-CD4 Mouse Antibody (redFluor® 710) [clone: OKT4]
Supplier: Tonbo Biosciences
80-0048-T100 80-0048-T025
10141-080EA 78.1 USD
10141-080 10141-252
Anti-CD4 Mouse Antibody (redFluor® 710) [clone: OKT4]
Antibodies
The OKT4 antibody reacts with human CD4, a 59 kDa protein which acts as a co-receptor for the T cell receptor (TCR) in its interaction with MHC Class II molecules on antigen-presenting cells. The extracellular domain of CD4 binds to the beta-2 domain of MHC Class II, while its cytoplasmic tail provides a binding site for the tyrosine kinase lck, facilitating the signaling cascade that initiates T cell activation. CD4, and co-receptors CCR5 and CXCR4, may also be utilized by HIV-1 to enter T cells. Human CD4 is typically expressed on thymocytes, some mature T cell populations such as Th17 andMore Product Information