Labels 'Dangerous When Wet', Labelmaster
Supplier: Label Master
HML23
101376-372PK 58.71 USD
101376-372
Labels 'Dangerous When Wet', Labelmaster
Labels
Employ a fool-proof shipping label solution with Worded Dangerous When Wet labels, revised to meet the design requirements of 49 CFR 172.407.
  • Printed with light-fast inks for fade resistance
  • Adhesives are formulated for use in various climates and environments
  • Once affixed, labels won't fade or fall off
  • Outstanding chemical and abrasion resistance
In addition to complying with directives set by the DOT, these labels also meet the design and durability standards of 49 CFR, ICAO, and other international air regulations.