10X PBS with 0.5% Tween 20, Teknova
Supplier: TEKNOVA INC
P0197
101320-452EA 136.65 USD
101320-452
10X PBS with 0.5% Tween 20, Teknova
Buffers Cell Culture Buffers
Tween 20 is a frequently used member of the polysorbate family of detergents.
  • Sterile
  • Shipping at ambient conditions
  • Store at room temperature
Contains 1.37 M sodium chloride (NaCl); 27 mM potassium chloride (KCl); 79.4 mM disodium phosphate (Na2PO4); 20.6 mM monopotassium phosphate (KH2PO4) and 0.5% (v/v) Tween-20 (polysorbate 20).