Copper Torch Ignitor Tape for Optima™ 8x00, PerkinElmer
Supplier: PerkinElmer
N0781097
10130-472EA 208.29 USD
10130-472
Copper Torch Ignitor Tape for Optima™ 8x00, PerkinElmer
Tapes
This copper foil torch ignitor tape is used with Optima™ 8x00 ICP-OES series instruments and can be used for both Optima™ and HybridXLT™ torches.
  • Roll of tape includes 50 pieces
This copper ignitor tape is also included in the Scott/Cross-Flow II Sample Introduction Kit (N0780616) and the Cyclonic/Concentric Sample Introduction Kit (N0780617) for Optima™ 8x00.