Rotary Evaporator Enclosures, Heidolph®
Supplier: Heidolph NA, LLC
035100210 035100200 035100230
10129-154EA 7757.3 USD
10129-154 10129-156 10129-158
Rotary Evaporator Enclosures, Heidolph®
Fume Hoods and Enclosures Extraction Cabinets
User-friendly safety enclosure for use with Rotary Evaporators
These system are engineered specifically for enclosing rotary evaporators, chromatography equipment, and robotic equipment while providing unrestricted access for operator interaction.
Clear polycarbonate shell and horizontal sliding doors provide visibility for monitoring the equipment, while the AirSafe™ automatic safety controller maintains airflow and monitors filter condition. At only 61cm (24") deep and 96.5cm (38") wide, this enclosure takes up very little counter space.
Operator exposure to chemical fumes andMore Product Information