Nalgene™ Graduated Cylinder Variety Pack, Thermo Scientific
Supplier: Thermo Fisher Scientific
3662-1234
10124-796CS 249.88 USD
10124-796
Nalgene™ Graduated Cylinder Variety Pack, Thermo Scientific
Cylinders Measuring Cylinders
A convenient and cost effective way to get started setting up a new lab, assembling student lab kits, or refreshing an existing lab.
  • Large blue octagonal base prevents rolling and tipping
  • Molded-in graduations are silk-screened for easy reading
  • Excellent chemical resistance for a wide variety of solutions
  • Impact resistant at temperatures above −40 °C
  • Autoclavable, graduated
  • Meets ASTM Class B, E1272 Class B and 21 CFR Part 177,1520 for food/beverage use
All sizes have a generous pour spout and molded-in graduations that are inked for easy reading. NoMore Product Information