Disposable Plastic Serological Pipettes, Argos Technologies
Supplier: Antyila Scientific
13002-14 13002-15 13002-16 13002-17 13002-20 13002-19 04395-22 04395-08 13002-26 04395-10 13002-24 04395-17 04395-19 04395-13 04395-14 04395-15 04395-16 04395-09 04395-20 04395-18 13002-28 13002-29 13002-30 13002-32 13002-37 04395-21 13002-12
89126-848CS 512.8 USD
89126-848 89126-850 89126-852 89126-854 89126-858 89126-856 89236-678 10124-792 10124-800 10124-804 10124-806 10124-810 10124-820 10124-822 10124-824 10124-826 10124-828 10124-830 10124-832 10124-834 10124-836 10124-838 10124-840 10124-842 10124-844 10124-846 10662-288 75805-148
Disposable Plastic Serological Pipettes, Argos Technologies
Pipettes Serological Pipettes
Argos brand standard serological pipettes are designed for accurate and rapid fluid dispensing
Argos Aspirating pipettes are sterile, individually wrapped plastic serological pipettes that are supplied without filter plugs and graduations for vacuum aspirating applications. They are very useful in tissue culture applications when aspirating larger volumes from dishes, flasks, or roller bottles.
Argos open end pipettes are just like the standard individually wrapped pipettes without the tip. The wide end opening is ideal when pipetting thick or viscous liguids, liquids that containMore Product Information