VWR® Solvent Waste System
Supplier: VWR International
10124-040 10124-042 10124-044 10124-342
10124-040EA 788.56 USD
10124-040 10124-042 10124-044 10124-342
VWR® Solvent Waste System
Safety Cans
This solvent waste system is designed to be an affordable solution for HPLC liquid waste disposal.
  • Activated carbon filter traps volatile organic compounds
  • Prevents chemicals from being inhaled
  • Closed-loop design
Each system is manufactured to safely collect waste from two HPLC machines.
This system features an activated carbon filter which traps volatile organic compounds from being released into the air. This prevents chemicals from being inhaled by lab workers and provides an easy mechanism of disposal for vapor wastes. The solvent waste systemMore Product Information