VWR® Cryogenic Dot and Strip Labels
Supplier: VWR International
89097-572 89097-574 89097-576 89097-578 89097-580 89097-582 89097-584 89097-586 89097-590 89097-592 89097-594 89097-596 89097-598 89097-600 89097-602 89097-604 89097-606 89097-608 89097-610 89097-612 89097-614 89097-616 89129-280 89129-282 89129-284 89129-286 89129-288 89129-290 89129-292 89129-294 89097-588 10123-758 10123-760 10123-762 10123-764 10123-766 10123-768 10123-770 10123-772 10123-774 10123-776 10123-778 10123-780 10123-782 10123-784 10123-786 10123-788 10123-790 10123-792 10123-794 10123-796 10123-798 10123-800 10123-802 10123-804 10123-806 10123-808 10123-810 10123-812 10123-814 10123-816 10123-818 10123-820 10123-822 10123-824 10123-826 10123-828 10123-830 10123-832 10123-834 10123-836 10123-838 10123-840 10123-842 10123-844 10123-846 10123-848 10123-852 10123-854 10123-856 10123-858 10123-860 10123-862 10123-864 10123-866 10123-868 10123-870 10123-872 10123-874
89097-572CS 51.68 USD
89097-572 89097-574 89097-576 89097-578 89097-580 89097-582 89097-584 89097-586 89097-590 89097-592 89097-594 89097-596 89097-598 89097-600 89097-602 89097-604 89097-606 89097-608 89097-610 89097-612 89097-614 89097-616 89129-280 89129-282 89129-284 89129-286 89129-288 89129-290 89129-292 89129-294 89097-588 10123-758 10123-760 10123-762 10123-764 10123-766 10123-768 10123-770 10123-772 10123-774 10123-776 10123-778 10123-780 10123-782 10123-784 10123-786 10123-788 10123-790 10123-792 10123-794 10123-796 10123-798 10123-800 10123-802 10123-804 10123-806 10123-808 10123-810 10123-812 10123-814 10123-816 10123-818 10123-820 10123-822 10123-824 10123-826 10123-828 10123-830 10123-832 10123-834 10123-836 10123-838 10123-840 10123-842 10123-844 10123-846 10123-848 10123-852 10123-854 10123-856 10123-858 10123-860 10123-862 10123-864 10123-866 10123-868 10123-870 10123-872 10123-874
VWR® Cryogenic Dot and Strip Labels
Labels
VWR® Cryogenic Dot and Strip Labels are ideal for all cryostorage, liquid nitrogen, refrigeration, and ultra-low temperature freezer applications.
  • Labels can be placed in most incubators
Labels feature a unique adhesive that can withstand extreme temperatures from –196...121 °C (–321...250 °F), without the need for lamination.
Ordering information: Rolls of labels are supplied on a 44 mm Ø (1³/₄") core. Laser printer label sheets are 21.6W×27.9L cm (8¹/₂×11"). Assorted labels come in blue, green, purple, red, and yellow. Roll of assorted labelsMore Product Information
Order Now
SPECIFICATIONS
Environmentally Preferable