Ammonium iron (II) sulfate 0.1 N

Supplier: AQUA SOLUTIONS MS

3840-1L 3840-500ML 3840-100ML 3840-4L
101226-510EA 58.36 USD
101226-510 101225-968 101226-914 101224-260
Ammonium iron (II) sulfate 0.1 N
Haematoxylin Harris

Certifications: Traceable to NIST.

Formula: (NH₄)₂Fe(SO₄)₂·6H₂O
MW: 392.14 g/mol
Density: 1.04213 g/mL
MDL Number: MFCD00150530
CAS Number: 7783-85-9
UN: 1760
ADR: 8 II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR