Sulfuric acid 25% (v/v) in aqueous solution

Supplier: AQUA SOLUTIONS MS
Danger

9006-1L 9006-20L 9006-4L
101223-362EA 41.73 USD
101223-362 101225-720 101224-590
Sulfuric acid 25% (v/v) in aqueous solution
Sulfuric acid
Formula: H₂SO₄
MDL Number: MFCD00064589
CAS Number: 7664-93-9
UN: 2796
ADR: 8,II

Order Now

Specification Test Results

Assay 25.00 ± 1.25 %
Appearance and Color Clear water white liquid


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR