Water, deionized ASTM Type II

Supplier: AQUA SOLUTIONS MS

Synonyms: Dihydrogen oxide

9765-10L 9765-20L 9765-4L 9765-15GAL 9765-1L 9765-500ML
101224-284EA 55.91 USD
101224-284 101224-618 101223-948 101225-152 101223-270 101227-004
Water, deionized ASTM Type II
Water
Formula: H₂O
MW: 18.02 g/mol
Boiling Pt: 100
Melting Pt: 0
MDL Number: MFCD00011332
CAS Number: 7732-18-5
Merck Index: 13,10098

Order Now

Specification Test Results

Prepared to formulation on file Yes
Meets ASTM Type II spec. Yes
Silica ≤ 3 ppb
Heterotropic plate count Zero
TOC ≤ 50 ppb
Sodium ≤ 5 ppb
Chloride ≤ 5 ppb
Conductivity ≤ 1.0 µMHOS
Heavy metals (as Pb) ≤ 10 ppb
Nitrate ≤ 0.4 ppm
Phosphate (PO₄) ≤ 1 ppm
Sulfate (SO₄) ≤ 1 ppm


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR