Toluene 99,5 ACS

Supplier: AQUA SOLUTIONS MS

Synonyms: Methylbenzene, Methacide, Benzene, methyl, Phenylmethane, Methane, phenyl-, Toluol, FM approval, Methylbenzol

T7601-20L T7601-500ML T7601-1L T7601-4L T7601-4LPVC
101225-286EA 364.02 USD
101225-286 101223-276 101223-656 101223-786 101224-814
Toluene 99,5 ACS
Toluene
Formula: C₆H₅CH₃
MW: 92.14 g/mol
Boiling Pt: 110.6
Melting Pt: -95
Flash Pt: 4
MDL Number: MFCD00008512
CAS Number: 108-88-3
UN: 1294
Merck Index: 13,09607
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=44efa31292a7dce6d9e60bcbaec208eb42d34e32&vid=73ada9ad1e894808b6c193cabc4babcfaf320a25&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Assay 99.5%
Color Clear
Form Liquid
Infrared Spectrum Conforms
Residue after Evaporation Max. 0.003%
Substances Darkened by Permanganate Passes Test
Sulfur Compounds as (S) Max 0.003%
Water (H₂O) Max. 0.03%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR