Potassium hydroxide 0.1 N isopropanol

Supplier: AQUA SOLUTIONS MS
Danger

Synonyms: Potash caustic, Potassium hydrate, Caustic potash

7241-1L 7241-500ML 7241-4L 7241-250ML
101202-900EA 67.87 USD
101202-900 101204-922 101203-974 103194-122
Potassium hydroxide 0.1 N isopropanol
Potassium hydroxide
Formula: KOH
MW: 56.11 g/mol
Boiling Pt: 1320
Melting Pt: 360
Storage Temperature: Ambient
MDL Number: MFCD00003553
CAS Number: 1310-58-3
UN: 1993
ADR: 3 II
Merck Index: 12,07806

Order Now

Specification Test Results

Normality 0.0995 - 0.1005


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR