Amersham™ Protran® Western Blotting Membranes, Nitrocellulose, Cytiva
Supplier: Cytiva
10600002 10600005 10600006 10600000 10600001 10600032 10600033 10600041 10600042 10600043 10600044 10600045 10600046 10600062 10600114 10600115 10600116 10600007 10600010 10600011 10600012 10600073 10600093 10600094 10600124 10600125
10063-173PK 577.1 USD
10063-173 10119-998 10120-000 10120-002 10120-004 10120-030 10120-032 10120-046 10120-048 10120-050 10120-052 10120-054 10120-056 10120-082 10120-130 10120-132 10120-134 10120-148 10120-162 10120-164 10120-166 10120-174 10120-180 10120-182 10147-176 10147-178
Amersham™ Protran® Western Blotting Membranes, Nitrocellulose, Cytiva
Transfer Membranes
Amersham™ Protran® membranes are manufactured from 100% pure nitrocellulose to ensure the highest binding capacity possible. They are compatible with a variety of detection methods, including isotopic, chemiluminescent, colorimetric and fluorescent. No methanol pre-wetting step is required, making this the membrane of choice for proteins which prefer aqueous environments. Prior to transfer the membrane is simply wetted with water, and then placed in the transfer buffer. No other pretreatment steps are necessary.
  • Proprietary nitrocellulose formula gives a long shelf life of bound
More Product Information