GC Syringes for Thermo Scientific TriPlus RSH Autosampler
Supplier: Thermo Scientific
365C6610 365C0221 365C0231 365C0241 365C0251 365D0261 365D0271 365D0281 365D0301 365D0311 365D0321 365D0331 365F2441 365F2461 365G2311 365G2321 365G2331 365H2141 365H2151 365H2161 365H2171 365H2181 365I2321 365I2331 365I2341 365I2351 365J2411 365J2421 365J2441 365K2811 365A0241 365B0251 365Q2121 365Q2131 365Q2141 365N2721
10057-472EA 166.04 USD
10057-472 10117-904 10117-906 10117-908 10117-910 10117-912 10117-914 10117-916 10117-920 10117-922 10117-924 10117-926 10117-930 10117-932 10117-934 10117-936 10117-938 10117-942 10117-944 10117-946 10117-948 10117-950 10117-952 10117-954 10117-956 10117-958 10117-960 10117-962 10117-964 10117-968 10117-970 10117-974 10117-976 10117-978 10117-980 10117-982 10117-984
GC Syringes for Thermo Scientific TriPlus RSH Autosampler
Syringes Syringes for Chromatography
Thermo Scientific™ syringes for GC instruments are designed to provide enhanced durability, clarity and accuracy in sample introduction. This gives confidence in analytical results, time-after-time.
  • Improved solvent resistance
  • Greater temperature range
  • Increased operational smoothness
  • Longer lifetime and improved cycle life
  • Closer fit between the plunger tip and PTFE insert at the zero position
  • Tighter fit between PTFE insert and glass barrel
  • Improved fixed needle attachment
Syringes are provided with a clear, easy-to-read black scale, which aids inMore Product Information