Legato 185 Single Syringe Pico Flow Infuse/Withdraw Programmable Syringe Pump, KD Scientific
Supplier: KD Scientific
788180SGL
10117-504EA 4511.88 USD
10117-504
Legato 185 Single Syringe Pico Flow Infuse/Withdraw Programmable Syringe Pump, KD Scientific
Pumps Syringe Pumps
Infuse/Withdraw/Continuos Single Picoliter syringe pump
Accomodates syringe sizes 0.5ul to 60ml. User definable flow rates. High resolutuion touch screen display. Programmable with two user defined programs. Selectable syringe table. Up to 30 lbs of linear force. USB control.
Certifications: UL, CSA, CUL, CE (Global), RoHS, WEEE.
Caution: KD Scientific syringe pumps are for laboratory use only. Not for clinical use.