Legato 111 Dual Syringe Infuse/Withdraw Programmable Syringe Pump, KD Scientific
Supplier: KD Scientific
788111
10117-492EA 4427.96 USD
10117-492
Legato 111 Dual Syringe Infuse/Withdraw Programmable Syringe Pump, KD Scientific
Pumps Syringe Pumps
Infuse/Withdraw/Continuous Dual Syringe pump
  • Touch pad lock feature, LED light on front panel, USB port, I/O and TTL interface
  • Full metal chassis
  • Built in syringe table, RS485 interface
  • Advanced microstepping techniques
  • Continuous mode of operation
  • Spill dam
  • Programmable
Accomodates syringe sizes 0.5ul to 10ml. User definable flow rates. High resolutuion touch screen display. Programmable with two user defined programs. Selectable syringe table. Up to 30 lbs of linear force. USB & RS232 control.
Certifications: UL, CSA, CUL, CE (Global), RoHS,More Product Information