Anti-DHX58 Goat Polyclonal Antibody

Supplier: Prosci
46-863
10113-108EA 715.98 USD
10113-108
Anti-DHX58 Goat Polyclonal Antibody
Antibodies
ELISA: Antibody detection limit dilution 1:8,000. Western Blot: Approximately 80 kDa band observed in human liver lysates (calculated MW of 76.6 kDa according to NP_077024.2). Recommended concentration: 0.3-1 ?g/mL.

LGP2 antibody is supplied in Tris saline, 0.02% sodium azide, pH 7.3 with 0.5% BSA.

Aliquot and store at -20°C. Avoid multiple freeze-thaw cycles.

Type: Primary
Antigen: DHX58
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR