Dextran, M.W. ~ 70,000 calibration_standard

Supplier: USP

1179887
101094-774EA 481.54 USD
101094-774
Dextran, M.W. ~ 70,000 calibration_standard
Dextran
Formula: (C₆H₁₀O₅)n
MDL Number: MFCD00130935
CAS Number: 9004-54-0
EINECS: 232-677-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR