Oakton T100 Replacement Turbidity Calibration Kit, Cole-Parmer
Supplier: OAKTON INSTRUMENTS MS
WD-35635-52
101076-390EA 642.92 USD
101076-390
Oakton T100 Replacement Turbidity Calibration Kit, Cole-Parmer
Turbidity Meters
For calibration of Oakton T-100 Turbidimeter.
Ordering information: Includes one each of 60 mL HPDE bottle of four NTU standards: 0.02, 20.0, 100 and 800 NTU.