Anti-Mycoplasma pneumoniae Rabbit Polyclonal Antibody

Supplier: Prosci
35-317
10071-990EA 587.64 USD
10071-990
Anti-Mycoplasma pneumoniae Rabbit Polyclonal Antibody
Antibodies
Purified IgG fraction of antiserum to a purity of >90%.

Mycoplasma pneumoniae antibody is in a phosphate saline buffer (0.01M, pH 7.2) containing 0.1% sodium azide preservative. No stabilizing proteins have been added.

Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.

Type: Primary
Antigen: Mycoplasma pneumoniae Antibody
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Bacteria
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR