ACTERO™ Salmonella Enrichment Media, FoodChek Systems
Supplier: FOODCHEK SYSTEMS INC.
FCM-009 FCM-088 FCM-089 FCM-090 FCM-049 FCM-064 FCM-065 FCM-066 FCM-085 FCM-086 FCM-087 FCM-050 FCM-051 FCM-140 FCM-141 FCM-142 FCM-143 FCM-144 FCM-145 FCM-146 FCM-147 FCM-148 FCM-188 FCM-189 FCM-190 FCM-107 FCM-109 FCLM-033
97068-836EA 1074.42 USD
97068-836 10066-030 10066-032 10066-034 10066-054 10066-056 10066-058 10066-060 10066-074 10066-076 10066-078 10066-108 10066-110 10146-700 10146-702 10146-704 10146-706 10146-708 10146-710 10146-712 10146-714 10146-716 10158-922 10158-924 10158-926 10500-800 10500-804 10500-812
ACTERO™ Salmonella Enrichment Media, FoodChek Systems
Media for Microbiology Dehydrated Media for Microbiology
Food and environmental samples require enrichment before pathogen testing enabling target pathogen present to grow and be detected in test samples
Using Actero™ Elite Enrichment Media significantly reduces all sample enrichment phase timelines regardless the type of "food pathogen testing system," such as lateral flow, ELISA and PCR platforms.
Actero™ Universal Enrichment Media are established formulations that are used in standard protocols employed in testing labs today.
The patented Actero™ Elite Enrichment Media are enhanced formulations that reduce the enrichment time required for accurate pathogen testing. Actero™ Salmonella Enrichement Media recieved the AOAC-PTM (041403) accreditation to be used as single-step recover and enrichment of Salmonella spp. from selected food and environmental samples.
Order Now
SPECIFICATIONS
Environmentally Preferable