Septa for Thermo TRACE PTV Inlets, Restek
Supplier: Restek
27094
10058-034PK 115.06 USD
10058-034 10058-036
Septa for Thermo TRACE PTV Inlets, Restek
Septa
Premium Restek non-stick BTO septa are preconditioned for easy ready-to-use condition upon arrival, and each batch is optimized for low bleed and temperature resistance.
  • Usable to 400 °C inlet temperature
  • New plasma coating eliminates sticking in the injection port
  • Precision molding ensures consistent, accurate fit
  • Partial predrilled CenterGuide design
  • Preconditioned and ready to use
  • Each batch GC-FID tested
  • Bleed and temperature optimized; ideal for demanding GC and GC-MS applications
These septa are designed to guarantee a consistent, accurate fitMore Product Information