Hardwood Locker Room Benches, Wisconsin Bench
Supplier: Wisconsin Bench (WB)
LBS9H024 LBS9H036 LBS9H048 LBS9H060 LBS9H072 LBS9H084 LBS9H096 LBS9H108 LBS9H120 LBS9H132 LBS9H144 LBS12024 LBS12036 LBS12048 LBS12060 LBS12072 LBS12084 LBS12096 LBS12120 LBS12144 LBS18024 LBS18036 LBS18042 LBS18048 LBS18060 LBS18072 LBS18096 LBS18120 LBS18144 LBS20042 LBS20048 LBS24024 LBS24036 LBS24042 LBS24048 LBS24060 LBS24072 LBS24096 LBS24120 LBS24144
10054-788EA 268.55 USD
10054-788 10054-790 10054-792 10054-794 10054-796 10054-798 10054-800 10054-802 10054-804 10054-806 10054-808 10054-810 10054-812 10054-814 10054-816 10054-818 10054-820 10054-822 10054-824 10054-826 10054-828 10054-830 10054-832 10054-834 10054-836 10054-838 10054-840 10054-842 10054-844 10054-846 10054-848 10054-850 10054-852 10054-854 10054-856 10054-858 10054-860 10054-862 10054-864 10054-866
Hardwood Locker Room Benches, Wisconsin Bench
Furniture Gowning Benches
1.25" thick hardwood finger-glued strips locker room bench with radius corners and edges for safety.
  • Two coats top and sides and one coat bottom clear catalyzed lacquer
  • Optional pedestals sold separately
The optional 16.25"H steel pedestals are in 14-gauge steel. Column pedestals have a 8.25" plate top and bottom with 2.375" center column. Trapezoid pedestals measure 15"W at the base and is recommended for bench widths of 12" or 18"
Ordering information: Maximum span between pedestals is 4 feet. Pedestals sold separately as optional individualMore Product Information