ASTM Approved Non-Mercury Tank Gauging Thermometers, Thermco
Supplier: Thermco
ACC272S1S ACC272S2S ACC272S3S ACC273S1S ACC273S2S ACC273S3S
10053-466EA 212.58 USD
10053-466 10053-468 10053-470 10053-472 10053-474 10053-476
ASTM Approved Non-Mercury Tank Gauging Thermometers, Thermco
Thermometers Analog Thermometers
Scoop Master model, a one piece molded black heat resistant plastic material impervious to the chemical action of petroleum products and aromatic hydrocarbons
Thermometers supplied with each case are ASTM approved, non-mercury blue spirit; remounted on spring clips and can be replaced if breakage occurs. Three ranges available in both celcius and farenheit.
The low thermal conductivity of the plastic maintains the temperatures of the sample. The case is non-sparking.
Certifications: ASTM approved/ISO17025/Statement of Traceability.
Ordering information: Cup Case Thermometer Only.More Product Information