Anti-CD86 Rat Monoclonal Antibody (PE (Phycoerythrin)) [clone: GL-1 (GL1)]
Supplier: Tonbo Biosciences
50-0862-U025 50-0862-U100
10051-282EA 49.5 USD
10051-282 10051-284
Anti-CD86 Rat Monoclonal Antibody (PE (Phycoerythrin)) [clone: GL-1 (GL1)]
Antibodies
The GL-1 antibody reacts with mouse CD86, also known as B7-2, an 80 kDa cell surface protein which is a ligand for CD28, a co-stimulatory receptor for the T cell receptor (TCR). CD28 can also bind a second B7 ligand known as CD80 (B7-1). Both CD80 and CD86 are expressed on activated B cells and antigen-presenting cells. These ligands trigger CD28 signaling in concert with TCR activation to drive T cell proliferation, induce high-level expression of IL-2, impart resistance to apoptosis, and enhance T cell cytotoxicity. The interaction / co-stimulatory signaling between the B7 ligands and CD28More Product Information