Anti-Static Spray, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
60615
100504-348EA 25.29 USD
100504-348
Anti-Static Spray, Electron Microscopy Sciences
Cleaning Agents Lens Cleaning Solutions
Anti static spray stops static build-up on optics, disc drives, photographic materials, lenses, metal rollers.