PTFE Beakers, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
60949-10C 60936-C 60938-C 60949-10 60949-20 60940-C 60949-06 60949-06C 60941-C 60938 60940 60947 60936 60947-C 60949-20C 60942 60944 60942-C 60948-C 60946 60944-C 60946-C 60941 60948
100497-484EA 40.39 USD
100497-484 100497-462 100497-464 100497-482 100497-486 100497-466 100497-478 100497-480 100501-622 100503-240 100503-282 100503-284 100503-294 100497-474 100497-488 100503-244 100503-246 100497-468 100497-476 100503-248 100497-470 100497-472 100503-242 100503-250
PTFE Beakers, Electron Microscopy Sciences
Beakers
These beakers are statically molded from virgin "PTFE" and machine finished to give a smooth surface for ease of cleaning and to eliminate contamination.
  • With pouring spout
  • Virtually immune to chemical attack
Working temperature range is from cryogenic to 260°C (500°F). Ideal for use in wafer etching.