Diamond Knife Cleaning Kit, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
71180 70970 70600 70610
100491-750EA 12.95 USD
100491-750 100500-628 100502-080 100496-272
Diamond Knife Cleaning Kit, Electron Microscopy Sciences
Wipes Cleaning Kits and Pads
This cleaning kit is used for the removal of sections from the knife edge and the trough.
Hair curl is made from human hair; good for removing sections, and cleaning the diamond knife.
Delivery information: Kit includes 5×5 ml vials of cleaning solution, 4 rods for cleaning the knife edge, and 1 hair curl.