Disposable Base Mold, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
62352-07 62352-30 62352-37 62352-24 62352-15
100501-994PK 157.82 USD
100501-994 100502-000 100502-002 100501-998 100501-996
Disposable Base Mold, Electron Microscopy Sciences
Moulds Moulds for Histology
Our Disposable Base Mold is designed to fit all existing cassettes. Because our base molds are disposable you save time on cleaning and storing and they offer you less exposure to hazardous chemicals.