Compartment - Micro Processing/Embedding Cassettes, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
70078-L 70078-P 70078-B 70078-GR 70078-A 70078-Y 70078-W 70078-PE 70078-O 70078-T 70078-G
100500-740BX 270.38 USD
100500-740 100500-744 100500-734 100500-738 100500-732 100500-752 100500-750 100500-746 100500-742 100500-748 100500-736
Compartment - Micro Processing/Embedding Cassettes, Electron Microscopy Sciences
Embedding Cassettes Biopsy Processing Cassettes
Made from Acetal Polymer. These biopsy cassettes can hold up to six tissue specimens, each one placed in it’s own compartment, numbered from 1 to 6 and measuring 7 x 12 x 5 mm (1/4 x 7/16 x 3/16"). Cover and base have over 3000 0.36 mm square openings. Cassettes keep specimen safely submerged in solvent and are totally resistant to the most histological solvents. The efficient flow-through slots maximize fluid exchange and insure proper drainage. The one-piece integral lid eliminates the need for separate steel lids. Large labeling areas are located on three sides of the cassette. TheMore Product Information