Beaker Buddy™ Rack, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
72365-01
100493-762EA 177.49 USD
100493-762
Beaker Buddy™ Rack, Electron Microscopy Sciences
Racks Tube Racks
Beaker Buddy™ racks are round, anodized aluminum rack for use with beakers, water baths and ice baths
Place the tubes in the rack, and then screw on the handle to hold down the cover. The handle is 51/8" long, and assist in the removal from hot beakers. Rack measures 3.5" diameter and holds up to 16 tubes.
These racks are ideal for use with 1.5 and 1.0 mL tubes. The rack has a thermometer slot for temperature monitoring and alimimnum legs for free-standing use.