Microwave Leakage Detector, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
72083-00
100493-612EA 444.37 USD
100493-612
Microwave Leakage Detector, Electron Microscopy Sciences
Electrical Meters Digital Multimeters
Evaluate the RF emissions of mobile phone, radio and TV transmission towers, wireless camera, Wi-Fi and LAN detection and installation, microwave oven leakage and other environmental safety concerns.
  • Display also indicates overload and low battery
Radio Frequency (RF) is the range of electromagnetic frequencies above the audio range and below infrared light. Features maximum and minimum hold, an audible alarm, and a zero adjustment to eliminate background EMF.
Dimensions: 130 x 56 x 38 mm. Weight: 170 g.
Delivery information: Comes with a softMore Product Information