Fixed Handle 55 Slide Rack, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
71422-FR
100493-320EA 251.5 USD
100493-320
Fixed Handle 55 Slide Rack, Electron Microscopy Sciences
Jars Staining Jars
A stainless steel rack with a fixed handle and grooves for 76×26 mm microscope slides held vertically for maximum use of space and solutions. Accommodates two vertical racks (#71421-VR), which totals 96 slides.