SafetySet, S.C.A.T.
Supplier: Avantor
10040-124EA 895.44 USD
10040-124 10040-126
SafetySet, S.C.A.T.
Closures Safety Caps and Safety Waste Caps
Solvent Safety Cap set suitable for use with all current HPLC systems
Set comes with four safety caps and all required connectors. Unused connectors can be closed by the blind screw delivered with the set. This makes the system universally applicable, even when less connectors are needed for a time.